siRNA Details
| Gene Symbol: 5430421F17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040352.1 | 3 | 717 | |||
| GenBank mRNA | AK017331.1 | - | 3 | 717 | ||
| AK142918.1 | - | 2 | 1549 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY717144.1 | - | 3 | 717 | Chr. 8 - 25304007 - 25306270 [-] on Build GRCm38 | |

5430421F17Rik
Genomic Map