siRNA Details
| Gene Symbol: 5430421F17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040352.1 | 3 | 1752 | |||
| GenBank mRNA | AK017331.1 | - | 3 | 1752 | ||
| AK142918.1 | - | 2 | 2584 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5430421F17Rik| Gene Symbol: 5430421F17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040352.1 | 3 | 1752 | |||
| GenBank mRNA | AK017331.1 | - | 3 | 1752 | ||
| AK142918.1 | - | 2 | 2584 | |||