siRNA Details
| Gene Symbol: Gm17501 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040843.1 | - | 1, 2 | 618 | ||
| FJ422280.1 | - | 1, 2 | 222 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY234475.1 | - | 1, 2 | 218 | Chr. 3 - 145650712 - 145651529 [+] on Build GRCm38 | |

Gm17501
Genomic Map