siRNA Details
| Gene Symbol: Gm17501 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040843.1 | - | 2 | 931 | ||
| FJ422280.1 | - | 2 | 535 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm17501| Gene Symbol: Gm17501 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040843.1 | - | 2 | 931 | ||
| FJ422280.1 | - | 2 | 535 | |||