siRNA Details
| Gene Symbol: Epb4.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019923.1 | - | 1 | 1691 | ||
| AK147339.1 | - | 1 | 2793 | |||
| AK148569.1 | - | 1 | 699 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epb4.1| Gene Symbol: Epb4.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019923.1 | - | 1 | 1691 | ||
| AK147339.1 | - | 1 | 2793 | |||
| AK148569.1 | - | 1 | 699 | |||