siRNA Details
| Gene Symbol: Epb4.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK148569.1 | - | 1 | 2335 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ042489.1 | - | 1 | 576 | Chr. 4 - 131934278 - 131935018 [-] on Build GRCm38 | |

Epb4.1
Genomic Map