siRNA Details
| Gene Symbol: Sirt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086910.1 | - | 1 | 2257 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB530585.2 | - | 1 | 372 | Chr. 7 - 140873841 - 140874468 [-] on Build GRCm38 | |

Sirt3
Genomic Map