siRNA Details
| Gene Symbol: 4930423K06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086441.1 | - | 1393 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB525750.2 | - | 1 | 230 | Chr. 15 - 89375078 - 89375708 [+] on Build GRCm38 | |

4930423K06Rik
Genomic Map