siRNA Details
| Gene Symbol: Gm11999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040327.1 | - | 2 | 168 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB635812.1 | - | 2 | 168 | Chr. 11 - 11642281 - 11685400 [-] on Build GRCm38 | |

Gm11999
Genomic Map