siRNA Details
| Gene Symbol: 5930403N24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031090.1 | - | 1 | 1129 | ||
| AK040320.1 | - | 1 | 773 | |||
| BC112909.1 | - | 1 | 1284 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_177177.1 | - | 1129 | Chr. - - [-] on Build | ||

5930403N24Rik
Genomic Map