siRNA Details
| Gene Symbol: 5430437J10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC023070.1 | - | 6 | 741 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI688362.1 | - | 6 | 743 | Chr. 15 - 5496266 - 5587760 [+] on Build GRCm38 | |

5430437J10Rik
Genomic Map