siRNA Details
| Gene Symbol: Gfm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020100.1 | - | 1 | 355 | ||
| AK082054.1 | - | 1 | 360 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gfm2| Gene Symbol: Gfm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020100.1 | - | 1 | 355 | ||
| AK082054.1 | - | 1 | 360 | |||