siRNA Details
| Gene Symbol: 6720407P12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020106.1 | - | 1 | 736 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA930303.1 | - | 1 | 40 | Chr. 11 - 69802135 - 69802241 [-] on Build GRCm38 | |

6720407P12Rik
Genomic Map