siRNA Details
| Gene Symbol: 5730408K05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011444.1 | - | 1 | 140 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY710716.1 | - | 1 | 140 | Chr. 19 - 8888389 - 8889144 [-] on Build GRCm38 | |

5730408K05Rik
Genomic Map