siRNA Details
| Gene Symbol: LOC100049077 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014010.1 | - | 1 | 459 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CB848666.2 | - | 1 | 467 | Chr. 9 - 21955293 - 21956005 [-] on Build GRCm38 | |

LOC100049077
Genomic Map