siRNA Details
| Gene Symbol: Egfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014017.1 | - | 1 | 917 | ||
| AK049452.1 | - | 1 | 809 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Egfr| Gene Symbol: Egfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014017.1 | - | 1 | 917 | ||
| AK049452.1 | - | 1 | 809 | |||