siRNA Details
| Gene Symbol: Hipk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003718.1 | - | 1 | 57 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY703977.1 | - | 1 | 57 | Chr. 6 - 38687899 - 38688786 [-] on Build GRCm38 | |

Hipk2
Genomic Map