siRNA Details
| Gene Symbol: 4933427E13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045853.1 | 6 | 978 | |||
| GenBank mRNA | AK016948.1 | - | 6 | 979 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933427E13Rik| Gene Symbol: 4933427E13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045853.1 | 6 | 978 | |||
| GenBank mRNA | AK016948.1 | - | 6 | 979 | ||