siRNA Details
Gene Symbol: 4933427E13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045853.1 | 7 | 1042 | |||
GenBank mRNA | AK016948.1 | - | 7 | 1043 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933427E13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045853.1 | 7 | 1042 | |||
GenBank mRNA | AK016948.1 | - | 7 | 1043 |