siRNA Details
| Gene Symbol: Gm8179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046039.1 | 1 | 316 | |||
| GenBank mRNA | AK143160.1 | - | 1 | 318 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY731383.1 | - | 1 | 318 | Chr. 8 - 32151893 - 32180882 [+] on Build GRCm38 | |

Gm8179
Genomic Map