siRNA Details
| Gene Symbol: Fam214a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087474.1 | - | 3 | 1933 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam214a| Gene Symbol: Fam214a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087474.1 | - | 3 | 1933 | ||