siRNA Details
| Gene Symbol: Gm29907 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082117.1 | - | 1583 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm29907| Gene Symbol: Gm29907 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082117.1 | - | 1583 | |||