siRNA Details
| Gene Symbol: A330093E20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040342.1 | 4 | 1985 | |||
| GenBank mRNA | AK039712.1 | - | 4 | 1905 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A330093E20Rik| Gene Symbol: A330093E20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040342.1 | 4 | 1985 | |||
| GenBank mRNA | AK039712.1 | - | 4 | 1905 | ||