siRNA Details
| Gene Symbol: Linc-md1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031267.1 | - | 1 | 3151 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Linc-md1| Gene Symbol: Linc-md1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031267.1 | - | 1 | 3151 | ||