siRNA Details
| Gene Symbol: 4930511M18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015762.1 | - | 4 | 413 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715304.1 | - | 4 | 413 | Chr. 3 - 92102573 - 92142736 [-] on Build GRCm38 | |

4930511M18Rik
Genomic Map