siRNA Details
| Gene Symbol: 4930532J02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015945.1 | - | 1 | 427 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715510.1 | - | 1 | 427 | Chr. 3 - 94953308 - 94953932 [-] on Build GRCm38 | |

4930532J02Rik
Genomic Map