siRNA Details
| Gene Symbol: 4930532J02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015945.1 | - | 1 | 550 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930532J02Rik| Gene Symbol: 4930532J02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015945.1 | - | 1 | 550 | ||