siRNA Details
| Gene Symbol: 4930532I03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015944.1 | - | 7 | 1396 | ||
| Gene Symbol: 2810408B13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013043.1 | - | 260 | |||

4930532I03Rik
Genomic Map