siRNA Details
| Gene Symbol: 4930532I03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015944.1 | - | 7 | 1178 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ236039.1 | - | 4 | 674 | Chr. 10 - 106555720 - 106601050 [-] on Build GRCm38 | |

4930532I03Rik
Genomic Map