siRNA Details
| Gene Symbol: 4930511P09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015767.1 | - | 1 | 58 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715311.1 | - | 1 | 55 | Chr. 18 - 56418378 - 56419111 [-] on Build GRCm38 | |

4930511P09Rik
Genomic Map