siRNA Details
| Gene Symbol: B230206I08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020990.1 | - | 1 | 175 | ||
| AK043775.1 | - | 1 | 176 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB641553.1 | - | 1 | 176 | Chr. 7 - 92410404 - 92411046 [+] on Build GRCm38 | |

B230206I08Rik
Genomic Map