siRNA Details
| Gene Symbol: 2900024J01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013590.1 | - | 1 | 1026 | ||
| AV151863.1 | - | 1 | 245 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2900024J01Rik| Gene Symbol: 2900024J01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013590.1 | - | 1 | 1026 | ||
| AV151863.1 | - | 1 | 245 | |||