siRNA Details
| Gene Symbol: 2900024J01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013590.1 | - | 1 | 395 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712550.1 | - | 1 | 386 | Chr. 1 - 180777850 - 180778762 [+] on Build GRCm38 | |

2900024J01Rik
Genomic Map