siRNA Details
| Gene Symbol: Gm15816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042176.1 | - | 6 | 1581 | ||
| BY612517.1 | - | 1 | 283 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15816| Gene Symbol: Gm15816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042176.1 | - | 6 | 1581 | ||
| BY612517.1 | - | 1 | 283 | |||