siRNA Details
| Gene Symbol: C030017B01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021087.1 | - | 1 | 743 | ||
| AK046738.1 | - | 1 | 554 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C030017B01Rik| Gene Symbol: C030017B01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021087.1 | - | 1 | 743 | ||
| AK046738.1 | - | 1 | 554 | |||