siRNA Details
Gene Symbol: C030017B01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021087.1 | - | 1 | 62 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728176.1 | - | 1 | 60 | Chr. 18 - 40539788 - 40540483 [+] on Build GRCm38 |
