siRNA Details
| Gene Symbol: C030014O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021082.1 | - | 1 | 279 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY728165.1 | - | 1 | 276 | Chr. 2 - 130840195 - 130840871 [+] on Build GRCm38 | |

C030014O09Rik
Genomic Map