siRNA Details
| Gene Symbol: 6330403N20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018109.1 | - | 1 | 180 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY718263.1 | - | 1 | 180 | Chr. 7 - 80298730 - 80299616 [-] on Build GRCm38 | |

6330403N20Rik
Genomic Map