siRNA Details
| Gene Symbol: 8430434A19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018454.1 | - | 1 | 993 | ||
| AV169955.1 | - | 1 | 179 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

8430434A19Rik| Gene Symbol: 8430434A19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018454.1 | - | 1 | 993 | ||
| AV169955.1 | - | 1 | 179 | |||