siRNA Details
| Gene Symbol: 8430429K09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045275.1 | 2 | 1632 | |||
| NR_045276.1 | 2 | 1791 | ||||
| GenBank mRNA | AK018450.1 | - | 2 | 1632 | ||
| BY645620.1 | - | 1 | 360 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

8430429K09Rik| Gene Symbol: 8430429K09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045275.1 | 2 | 1632 | |||
| NR_045276.1 | 2 | 1791 | ||||
| GenBank mRNA | AK018450.1 | - | 2 | 1632 | ||
| BY645620.1 | - | 1 | 360 | |||