siRNA Details
Gene Symbol: 8430429K09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045275.1 | 2 | 1298 | |||
NR_045276.1 | 2 | 1457 | ||||
GenBank mRNA | AK018450.1 | - | 2 | 1298 | ||
BY645620.1 | - | 1 | 26 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 8430429K09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045275.1 | 2 | 1298 | |||
NR_045276.1 | 2 | 1457 | ||||
GenBank mRNA | AK018450.1 | - | 2 | 1298 | ||
BY645620.1 | - | 1 | 26 |