siRNA Details
| Gene Symbol: Gm16938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045969.1 | 4 | 2347 | |||
| NR_045970.1 | 4 | 2323 | ||||
| NR_045971.1 | 4 | 2320 | ||||
| NR_045972.1 | 3 | 2211 | ||||
| GenBank mRNA | AK085722.1 | - | 2213 | |||
| BC030866.1 | - | 1 | 1449 | |||
| BC040816.1 | - | 1 | 1437 | |||

Gm16938
Genomic Map