siRNA Details
| Gene Symbol: Gm3693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046721.1 | - | 79 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB594062.2 | - | 1 | 79 | Chr. 12 - 81228284 - 81241043 [+] on Build GRCm38 | |

Gm3693
Genomic Map