siRNA Details
| Gene Symbol: Gm4278 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046078.1 | 3 | 316 | |||
| GenBank mRNA | AK139702.1 | - | 3 | 317 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY725599.1 | - | 3 | 317 | Chr. 14 - 74975086 - 74986316 [+] on Build GRCm38 | |

Gm4278
Genomic Map