siRNA Details
Gene Symbol: Gm4278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046078.1 | 3 | 458 | |||
GenBank mRNA | AK139702.1 | - | 3 | 459 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY725599.1 | - | 3 | 459 | Chr. 14 - 74975086 - 74986316 [+] on Build GRCm38 |
