siRNA Details
| Gene Symbol: Gm4278 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046078.1 | 3 | 1018 | |||
| GenBank mRNA | AK139702.1 | - | 3 | 1019 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4278| Gene Symbol: Gm4278 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046078.1 | 3 | 1018 | |||
| GenBank mRNA | AK139702.1 | - | 3 | 1019 | ||