siRNA Details
| Gene Symbol: 4933439N14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019859.1 | - | 2 | 377 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716808.1 | - | 2 | 377 | Chr. 8 - 12502899 - 12504838 [+] on Build GRCm38 | |

4933439N14Rik
Genomic Map