siRNA Details
| Gene Symbol: 4930509G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006093.1 | - | 1 | 79 | ||
| AK015735.1 | - | 1 | 1327 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930509G22Rik| Gene Symbol: 4930509G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006093.1 | - | 1 | 79 | ||
| AK015735.1 | - | 1 | 1327 | |||