siRNA Details
| Gene Symbol: 1700065J11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040526.1 | 3 | 353 | |||
| GenBank mRNA | AK006894.1 | - | 3 | 354 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706886.1 | - | 3 | 354 | Chr. 6 - 35330846 - 35336827 [+] on Build GRCm38 | |

1700065J11Rik
Genomic Map