siRNA Details
| Gene Symbol: 1700065J11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040526.1 | 2 | 89 | |||
| GenBank mRNA | AK006894.1 | - | 2 | 90 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706886.1 | - | 2 | 90 | Chr. 6 - 35330846 - 35336827 [+] on Build GRCm38 | |

1700065J11Rik
Genomic Map